Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-(methoxymethoxy)pyridine 96%

Supplier: Origen Biomedical, Inc
Warning

H52324-03 H52324-06
AAH52324-03EA 126 USD InStock
AAH52324-03 AAH52324-06 AAH52324-MD
2-Bromo-3-(methoxymethoxy)pyridine 96%
2-Bromo-3-(methoxymethoxy)pyridine
Purity: 96% CAS No.: 162271-10-5 Molecular Formula: C7H8BrNO2 Formula Weight: 218.05 Melting Point: 35-37° MDL No.: MFCD17015031


Formula: C₇H₈BrNO₂
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD17015031
CAS Number: 162271-10-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty