Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Carbamoylpyridine-3-boronic acid pinacol ester 96%

Supplier: Origen Biomedical, Inc
Warning

H52330-03 H52330-MD
AAH52330-03EA 506 USD InStock
AAH52330-03 AAH52330-MD
5-Carbamoylpyridine-3-boronic acid pinacol ester 96%
5-Carbamoylpyridine-3-boronic acid pinacol ester
Purity: 96% CAS No.: 1169402-51-0 Molecular Formula: C12H17BN2O3 Formula Weight: 248.09 MDL No.: MFCD11878274


Formula: C₁₂H₁₇BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11878274
CAS Number: 1169402-51-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty