Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1-Methyl-4-piperidinylmethoxy)aniline 96%

Supplier: Origen Biomedical, Inc
Warning

H52351-03 H52351-MD
AAH52351-03EA 469 USD InStock
AAH52351-03 AAH52351-MD
4-(1-Methyl-4-piperidinylmethoxy)aniline 96%
4-(1-Methyl-4-piperidinylmethoxy)aniline
Purity: 96% CAS No.: 902454-26-6 Molecular Formula: C13H20N2O Formula Weight: 220.31 Storage Temperature: 2°C to 8°C UN No.: 2811 MDL No.: MFCD09909628


Formula: C₁₃H₂₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD09909628
CAS Number: 902454-26-6
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty