Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methylsulphonyl)phenylboronic acid 98%

Supplier: Origen Biomedical, Inc
Warning

H52445-03 H52445-06
AAH52445-03EA 128 USD InStock
AAH52445-03 AAH52445-06
2-(Methylsulphonyl)phenylboronic acid 98%
2-(Methylsulphonyl)phenylboronic acid
Purity: 98% CAS No.: 330804-03-0 Molecular Formula: C7H9BO4S Formula Weight: 200.02 Melting Point: 78-84° MDL No.: MFCD02179472


Formula: C₇H₉BO₄S
Melting Pt: 78…84 °C
Storage Temperature: Ambient
MDL Number: MFCD02179472
CAS Number: 330804-03-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty