Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-pyridineboronic acid 95%

Supplier: Origen Biomedical, Inc
Warning

H52515-03 H52515-MD
AAH52515-03EA 204 USD InStock
AAH52515-03 AAH52515-MD
5-Chloro-2-pyridineboronic acid 95%
5-Chloro-2-pyridineboronic acid
Purity: 95% CAS No.: 652148-91-9 Molecular Formula: C5H5BClNO2 Formula Weight: 157.37 MDL No.: MFCD07368862


Formula: C₅H₅BClNO₂
MW: 157.36
Storage Temperature: Ambient
MDL Number: MFCD07368862
CAS Number: 652148-91-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty