Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-N-Boc-4-cyano-L-proline 97%

Supplier: Origen Biomedical, Inc
Warning

H52563-03 H52563-MD
AAH52563-03EA 1008 USD InStock
AAH52563-03 AAH52563-MD
trans-N-Boc-4-cyano-L-proline 97%
trans-N-Boc-4-cyano-L-proline
Purity: 97% CAS No.: 273221-94-6 Molecular Formula: C11H16N2O4 Formula Weight: 240.26 MDL No.: MFCD01860670


Formula: C₁₁H₁₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD01860670
CAS Number: 273221-94-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty