Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-trans-4-amino-L-proline methyl ester hydrochloride salt 97%

Supplier: Origen Biomedical, Inc

H52566-03 H52566-06
AAH52566-03EA 156 USD InStock
AAH52566-03 AAH52566-06 AAH52566-MD
Boc-trans-4-amino-L-proline methyl ester hydrochloride salt 97%
Boc-trans-4-amino-L-proline methyl ester hydrochloride salt
Purity: 97% CAS No.: 334999-32-5 Molecular Formula: C11H20N2O4.HCl Formula Weight: 280.75 MDL No.: MFCD03787929


Formula: C₁₁H₂₁ClN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD03787929
CAS Number: 334999-32-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty