Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid 97%

Supplier: Origen Biomedical, Inc

H52775-03 H52775-MD
AAH52775-03EA 685 USD InStock
AAH52775-03 AAH52775-MD
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid 97%
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid
Purity: 97% CAS No.: 62023-65-8 Molecular Formula: C15H21NO5 Formula Weight: 295.34 Beilstein Reference: 5909790 MDL No.: MFCD00057841


Formula: C₁₅H₂₁NO₅
MW: 295.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057841
CAS Number: 62023-65-8
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty