Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-4-amino-2-chlorobenzoic acid 97%

Supplier: Origen Biomedical, Inc
Warning

H52786-03 H52786-06 H52786-MD
AAH52786-03EA 178 USD InStock
AAH52786-03 AAH52786-06 AAH52786-MD
Boc-4-amino-2-chlorobenzoic acid 97%
Boc-4-amino-2-chlorobenzoic acid
Purity: 97% CAS No.: 232275-73-9 Molecular Formula: C12H14ClNO4 Formula Weight: 271.70 MDL No.: MFCD02682164


Formula: C₁₂H₁₄ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD02682164
CAS Number: 232275-73-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty