Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lovastatin 97%

Supplier: Origen Biomedical, Inc
Warning

H52792-06 H52792-14
AAH52792-06EA 120 USD InStock
AAH52792-06 AAH52792-14
Lovastatin 97%
Lovastatin
Purity: 97% CAS No.: 75330-75-5 Molecular Formula: C24H36O5 Formula Weight: 404.56 Melting Point: 175° MDL No.: MFCD00072164


Formula: C₂₄H₃₆O₅
MW: 404.55 g/mol
Melting Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00072164
CAS Number: 75330-75-5
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty