Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate 97%

Supplier: Origen Biomedical, Inc
Warning

H52832-03 H52832-06
AAH52832-03EA 57.3 USD InStock
AAH52832-03 AAH52832-06
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate 97%
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate
Purity: 97% CAS No.: 162167-97-7 Molecular Formula: C11H22N2O2 Formula Weight: 214.31 MDL No.: MFCD01317792


Formula: C₁₁H₂₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01317792
CAS Number: 162167-97-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty