H53109-03
H53109-MD
AAH53109-03EA
168
USD
InStock
AAH53109-03
AAH53109-MD
3-Fluoro-4-pyridineboronic acid 98%
3-Fluoro-4-pyridineboronic acid
Purity: 98%
CAS No.: 458532-97-3
Molecular Formula: C5H5BFNO2
Formula Weight: 140.91
Melting Point: 237-239°
MDL No.: MFCD03788558