H53406-03
H53406-06
H53406-14
AAH53406-03EA
34.2
USD
InStock
AAH53406-03
AAH53406-06
AAH53406-14
N-(tert-Butoxycarbonyl)propargylamine 97%
N-(tert-Butoxycarbonyl)propargylamine
Purity: 97%
CAS No.: 92136-39-5
Molecular Formula: C8H13NO2
Formula Weight: 155.20
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Melting Point: 40-44°
MDL No.: MFCD07367245