Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-pentyne 97%

Supplier: Origen Biomedical, Inc
Warning

H53433-06 H53433-14
AAH53433-06EA 76.1 USD InStock
AAH53433-06 AAH53433-14
1-Bromo-2-pentyne 97%
1-Bromo-2-pentyne
Purity: 97% CAS No.: 16400-32-1 Molecular Formula: C5H7Br Formula Weight: 147.01 Flash Point: 41°(106°F) Refractive Index: 1.498 Boiling Point: 93-94°/113mm UN No.: 1993 MDL No.: MFCD00236363


Formula: C₅H₇Br
Boiling Pt: 93…94 °C/113 mm
Density: 1.438
Flash Pt: 41 °C (106 °F)
Storage Temperature: Ambient
MDL Number: MFCD00236363
CAS Number: 16400-32-1
EINECS: 240-451-2
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty