Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl propargyl ether 98%

Supplier: Origen Biomedical, Inc
Danger

H53480-06 H53480-14 H53480-22
AAH53480-06EA 44.1 USD InStock
AAH53480-06 AAH53480-14 AAH53480-22
tert-Butyl propargyl ether 98%
tert-Butyl propargyl ether
Purity: 98% CAS No.: 16314-18-4 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 20°(68°F) Refractive Index: 1.418 Boiling Point: 119° UN No.: 1993 MDL No.: MFCD07368651


Formula: C₇H₁₂O
MW: 112.172 g/mol
Boiling Pt: 119 °C
Density: 0.835
Flash Pt: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD07368651
CAS Number: 16314-18-4
UN: 3271
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty