H53519-03
H53519-06
H53519-14
AAH53519-03EA
46.6
USD
InStock
AAH53519-03
AAH53519-06
AAH53519-14
6-Heptynoic acid 95%
6-Heptynoic acid
Purity: 95%
CAS No.: 30964-00-2
Molecular Formula: C7H10O2
Formula Weight: 126.16
Flash Point: 113°(235°F)
Refractive Index: 1.451
Boiling Point: 93-94°/1mm
Beilstein Reference: 1747024
UN No.: 3265
MDL No.: MFCD00239430