H54156-03
H54156-06
AAH54156-03EA
84
USD
InStock
AAH54156-03
AAH54156-06
AAH54156-MD
4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline 97%
4-[2-(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolinyl)ethyl]aniline
Purity: 97%
CAS No.: 82925-02-8
Molecular Formula: C19H24N2O2
Formula Weight: 312.41
MDL No.: MFCD04974539