H54283-03
H54283-06
H54283-MD
AAH54283-03EA
64.2
USD
InStock
AAH54283-03
AAH54283-06
AAH54283-MD
4-(Hydroxymethyl)benzonitrile 97%
4-(Hydroxymethyl)benzonitrile
Purity: 97%
CAS No.: 874-89-5
Molecular Formula: C8H7NO
Formula Weight: 133.15
Melting Point: 32-36°
UN No.: 3439
MDL No.: MFCD00870633