Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Pyridyl)benzimidazole-6-carboxylic acid 97%

Supplier: Origen Biomedical, Inc
Warning

H54593-03 H54593-06 H54593-MD
AAH54593-03EA 185.85 USD InStock
AAH54593-03 AAH54593-06 AAH54593-MD
2-(3-Pyridyl)benzimidazole-6-carboxylic acid 97%
2-(3-Pyridyl)benzimidazole-6-carboxylic acid
Purity: 97% CAS No.: 312496-16-5 Molecular Formula: C13H9N3O2 Formula Weight: 239.23 MDL No.: MFCD01414693


Formula: C₁₃H₉N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01414693
CAS Number: 312496-16-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty