H54704-03
H54704-06
H54704-MD
AAH54704-03EA
130
USD
InStock
AAH54704-03
AAH54704-06
AAH54704-MD
Methyl-2-amino-5-methoxybenzoate 96%
Methyl-2-amino-5-methoxybenzoate
Purity: 96%
CAS No.: 2475-80-1
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Melting Point: 32-33°
MDL No.: MFCD09038482