Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2,6-dimethylpyrimidine 97%

Supplier: Origen Biomedical, Inc
Warning

H54735-06 H54735-MD
AAH54735-03EA 160.95 USD InStock
AAH54735-03 AAH54735-06 AAH54735-MD
4-Chloro-2,6-dimethylpyrimidine 97%
4-Chloro-2,6-dimethylpyrimidine
Purity: 97% CAS No.: 4472-45-1 Molecular Formula: C6H7ClN2 Formula Weight: 142.03 MDL No.: MFCD00234197


Formula: C₆H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00234197
CAS Number: 4472-45-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty