H54831-03
H54831-06
H54831-MD
AAH54831-03EA
231
USD
InStock
AAH54831-03
AAH54831-06
AAH54831-MD
1-Amino-4-cyclopentylpiperazine 97%
1-Amino-4-cyclopentylpiperazine
Purity: 97%
CAS No.: 61379-64-4
Molecular Formula: C9H19N3
Formula Weight: 169.27
MDL No.: MFCD00190598