Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-4'-octyloxybiphenyl 97%

Supplier: Origen Biomedical, Inc
Warning

H55191-03 H55191-06
AAH55191-03EA 66.6 USD InStock
AAH55191-03 AAH55191-06
4-Cyano-4'-octyloxybiphenyl 97%
4-Cyano-4'-octyloxybiphenyl
Purity: 97% CAS No.: 52364-73-5 Molecular Formula: C21H25NO Formula Weight: 307.44 Flash Point: 113°(235°F) Melting Point: 51-77° MDL No.: MFCD00075145


Formula: C₂₁H₂₅NO
Melting Pt: 51…77 °C
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075145
CAS Number: 52364-73-5
EINECS: 257-878-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty