Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Bromophenyl)benzenesulfonamide 97%

Supplier: Origen Biomedical, Inc
Warning

H55476-03 H55476-MD
AAH55476-03EA 327 USD InStock
AAH55476-03 AAH55476-MD
N-(4-Bromophenyl)benzenesulfonamide 97%
N-(4-Bromophenyl)benzenesulphonamide
Purity: 97% CAS No.: 16468-97-6 Molecular Formula: C12H10BrNO2S Formula Weight: 312.18 MDL No.: MFCD00159375


Formula: C₁₂H₁₀BrNO₂S
Storage Temperature: Ambient
MDL Number: MFCD00159375
CAS Number: 16468-97-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty