Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyloxyaniline 98%

Supplier: Origen Biomedical, Inc
Warning

H55708-03 H55708-06 H55708-14
AAH55708-03EA 20.8 USD InStock
AAH55708-03 AAH55708-06 AAH55708-14
3-Benzyloxyaniline 98%
3-Benzyloxyaniline
Purity: 98% CAS No.: 1484-26-0 Molecular Formula: C13H13NO Formula Weight: 199.25 Physical Form: Solid Appearance: Pale brown Melting Point: 63-67° MDL No.: MFCD00007784


Formula: C₁₃H₁₃NO
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00007784
CAS Number: 1484-26-0
EINECS: 216-056-6
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty