Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicyclohexyl phthala 99+%

Supplier: Origen Biomedical, Inc
Danger

H56004-22 H56004-36
AAH56004-22EA 36.6 USD InStock
AAH56004-22 AAH56004-36
Dicyclohexyl phthala 99+%
Dicyclohexyl phthala
Purity: 99+% CAS No.: 84-61-7 Molecular Formula: C20H26O4 Formula Weight: 330.42 Refractive Index: 1.4310 Melting Point: 63-67° Beilstein Reference: 1889288 MDL No.: MFCD00003849


Formula: C₂₀H₂₆O₄
MW: 330.42 g/mol
Melting Pt: 63…67 °C
Density: 1.383
Storage Temperature: Ambient
MDL Number: MFCD00003849
CAS Number: 84-61-7
EINECS: 201-545-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty