Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cbz-Piperazine 98%

Supplier: Origen Biomedical, Inc
Warning

H56008-AA H56008-AC H56008-AE
AAH56008-AAEA 80 USD InStock
AAH56008-AA AAH56008-AC AAH56008-AE
1-Cbz-Piperazine 98%
1-Cbz-piperazine
Purity: 98% CAS No.: 31166-44-6 Molecular Formula: C12H16N2O2 Formula Weight: 220.27 Refractive Index: 1.5460 Boiling Point: 158-161°/1.4mm Beilstein Reference: 179500 MDL No.: MFCD00274317


Formula: C₁₂H₁₆N₂O₂
MW: 220.272 g/mol
Boiling Pt: 158…161 °C/1.4 mm
Density: 1.142
Storage Temperature: Ambient
MDL Number: MFCD00274317
CAS Number: 31166-44-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty