H56015-03
H56015-06
AAH56015-03EA
21.8
USD
InStock
AAH56015-03
AAH56015-06
AAH56015-14
2-Amino-3,5-dibromobenzaldehyde 97%
2-Amino-3,5-dibromobenzaldehyde
Purity: 97%
CAS No.: 50910-55-9
Molecular Formula: C7H5Br2NO
Formula Weight: 278.93
Melting Point: 130-135°
MDL No.: MFCD00671100