Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Diazafluoren-9-one 98%

Supplier: Origen Biomedical, Inc
Warning

H56028-03 H56028-06
AAH56028-03EA 134 USD InStock
AAH56028-03 AAH56028-06
4,5-Diazafluoren-9-one 98%
4,5-Diazafluoren-9-one
Purity: 98% CAS No.: 50890-67-0 Molecular Formula: C11H6N2O Formula Weight: 182.18 Melting Point: 214-217° MDL No.: MFCD00046892


Formula: C₁₁H₆N₂O
Melting Pt: 214…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00046892
CAS Number: 50890-67-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty