Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium phenylpyruvate 98%

Supplier: Origen Biomedical, Inc

H56767-06 H56767-14
AAH56767-06EA 74.24 USD InStock
AAH56767-06 AAH56767-14
Sodium phenylpyruvate 98%
Sodium phenylpyruvate
Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

Purity: 98% CAS No.: 114-76-1 Molecular Formula: C9H7NaO3 Formula Weight: 186.14 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 4770978 MDL No.: MFCD00002590


Formula: C₉H₇NaO₃
MW: 186.14 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002590
CAS Number: 114-76-1
EINECS: 204-053-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty