Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Phenylcarbazole 99%

Supplier: Origen Biomedical, Inc
Danger

H56827-06 H56827-14
AAH56827-06EA 150 USD InStock
AAH56827-06 AAH56827-14
9-Phenylcarbazole 99%
9-Phenylcarbazole
Purity: 99% CAS No.: 1150-62-5 Molecular Formula: C18H13N Formula Weight: 243.30 Melting Point: 95-97° MDL No.: MFCD00004965


Formula: C₁₈H₁₃N
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004965
CAS Number: 1150-62-5
EINECS: 214-564-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty