Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-Deprenyl hydrochloride

Supplier: Origen Biomedical, Inc
Warning

J61286-03 J61286-MD
AAJ61286-03EA 245 USD InStock
AAJ61286-03 10814-248
(R)-(-)-Deprenyl hydrochloride
(R)-(-)-Deprenyl hydrochloride
Selective MAO-B inhibitor; anti-Parkinsonian agent

Crystalline Powder

Applications: Selective MAO-B inhibitor; anti-Parkinsonian agent


Formula: C₁₃H₁₈ClN
MW: 223.744 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069299
CAS Number: 14611-52-0
Merck Index: 14,8428
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty