Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SB203580 98%

Supplier: Origen Biomedical, Inc
Danger

J61482-MB J61482-MC J61482-MF
AAJ61482-MBEA 153 USD InStock
AAJ61482-MB AAJ61482-MC AAJ61482-MF
SB203580 98%
SB203580


Formula: C₂₁H₁₆FN₃OS
Storage Temperature: Refrigerator
MDL Number: MFCD00922198
CAS Number: 152121-47-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty