Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bumetanide ≥98%

Supplier: Origen Biomedical, Inc

J62302-03 J62302-06
AAJ62302-03EA 131.56 USD InStock
AAJ62302-03 AAJ62302-06
Bumetanide ≥98%
Bumetanide
Inhibitor of the sodium-potassium-chloride cotransporter

Solid

Applications: Inhibitor of the sodium-potassium-chloride cotransporter


Formula: C₁₇H₂₀N₂O₅S
MW: 364.416 g/mol
Melting Pt: 230…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00078949
CAS Number: 28395-03-1
EINECS: 249-004-6
Merck Index: 14,1484
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty