Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Croton Oil

Supplier: Origen Biomedical, Inc
Danger

J62367-09 J62367-14 J62367-22
AAJ62367-09EA 61.7 USD InStock
AAJ62367-09 AAJ62367-14 AAJ62367-22
Croton Oil
Croton oil
A natural source of phorbol, phorbol esters and glycerides of saturated fatty acids

Oil

Applications: A natural source of phorbol, phorbol esters and glycerides of saturated fatty acids


Flash Pt: 74 °C (165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00130880
CAS Number: 8001-28-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty