Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibucaine hydrochloride

Supplier: Origen Biomedical, Inc
Danger

J62804-06 J62804-14
AAJ62804-06EA 63.4 USD InStock
AAJ62804-06 AAJ62804-14
Dibucaine hydrochloride
Dibucaine hydrochloride
Potent, long-acting local anesthetic

Crystalline powder

Applications: Potent, long-acting local anesthetic


Formula: C₂₀H₃₀ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00012735
CAS Number: 61-12-1
EINECS: 200-498-1
Merck Index: 14,3031
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty