Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bovine Hemoglobin

Supplier: Origen Biomedical, Inc

J63838-06 J63838-14 J63838-22
AAJ63838-06EA 41.06 USD InStock
AAJ63838-06 AAJ63838-14 AAJ63838-22
Bovine Hemoglobin
Proteins and Peptides
Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide.

  • Dark Red to Brown Lyophilized Powder, 64.5 kDa
  • Store at Cold Temperatures
  • Soluble in water

Used as a coloring agent for pet foods, a natural source of iron for nutraceuticals, a protein source for non-ruminant animals, and as a raw material for pharmaceutical porphyrin derivative production.

Precaución: Incompatible with oxidizng agents and heat.


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty