Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Apramycin sulfate 96%

Supplier: Origen Biomedical, Inc
Danger

J63874-03 J63874-06
AAJ63874-03EA 107.14 USD InStock
AAJ63874-03 AAJ63874-06
Apramycin sulfate 96%
Apramycin sulfate
Potent inhibitor of protein synthesis in bacteria

Powder

Applications: Potent inhibitor of protein synthesis in bacteria


Formula: C₂₁H₄₁N₅O₁₁·H₂SO₄
Boiling Pt: 823 °C
Melting Pt: 168 °C
Density: 1.56
Flash Pt: 451.6 °C(845 °F)
Storage Temperature: Ambient
MDL Number: MFCD06200257
CAS Number: 65710-07-8
EINECS: 265-890-7
Merck Index: 14,752
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty