Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloropurine 2'-deoxyriboside 97%

Supplier: Origen Biomedical, Inc
Warning

J64871-03 J64871-MD
AAJ64871-03EA 380 USD InStock
AAJ64871-03 10814-350
6-Chloropurine 2'-deoxyriboside 97%
6-Chloropurine 2'-deoxyriboside
Powder

Purity: 97% CAS No.: 4594-45-0 Molecular Formula: C10H11ClN4O3 Formula Weight: 270.67 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to off-white MDL No.: MFCD00083282


Formula: C₁₀H₁₁ClN₄O₃
MW: 270.673 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00083282
CAS Number: 4594-45-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty