Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Fluoro-2'-deoxyinosine 99%

Supplier: Origen Biomedical, Inc

J65208-03 J65208-MD
AAJ65208-03EA 689 USD InStock
AAJ65208-03 10814-378
2'-Fluoro-2'-deoxyinosine 99%
2'-Fluoro-2'-deoxyinosine
Powder

Purity: 99% CAS No.: 80049-87-2 Molecular Formula: C10H11FN4O4 Formula Weight: 270.22 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD09750860


Formula: C₁₀H₁₁FN₄O₄
Storage Temperature: Ambient
MDL Number: MFCD09750860
CAS Number: 80049-87-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty