Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylamine 40% (w/w) in aqueous solution

Supplier: Origen Biomedical, Inc
Danger

L00894-0F L00894-AK L00894-AU
AAL00894-0FEA 140.32 USD InStock
AAL00894-0F AAL00894-AK AAL00894-AU
Methylamine 40% (w/w) in aqueous solution
Methylamine
CAS No.: 74-89-5 Molecular Formula: CH3NH2 Formula Weight: 31.06 Flash Point: -14°(6°F) Refractive Index: 1.3710 Odor: Ammonia-like Boiling Point: 49° Beilstein Reference: 741851 Merck Reference: 14,6014 UN No.: 1235 MDL No.: MFCD00008104


Formula: CH₅N
MW: 31.06
Boiling Pt: 48
Melting Pt: -92.5
Density: 0.899
Flash Pt: –14 °C (6 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008104
CAS Number: 74-89-5
EINECS: 200-820-0
UN: 1235
ADR: 3 II
Merck Index: 14,6014
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty