L13278-0B
L13278-18
L13278-30
AAL13278-0BEA
123.67
USD
InStock
AAL13278-0B
AAL13278-18
AAL13278-30
Sodium cyanide 98%, granules
Sodium cyanide
Purity: 98%
CAS No.: 143-33-9
Molecular Formula: CNNa
Formula Weight: 49.01
Odor: schwach nach bitt.Mande
Melting Point: 563°
Boiling Point: ca 1500°
Beilstein Reference: 3587243
Merck Reference: 14,8605
UN No.: 1689
MDL No.: MFCD00003523