Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bovine Native Calmodulin (from Testes)

Supplier: Origen Biomedical, Inc

J60231-EXC J60231-MCR
AAJ60231-EXCEA 578 USD InStock
AAJ60231-EXC AAJ60231-MCR
Bovine Native Calmodulin (from Testes)
Proteins and Peptides
Lyophilized powder, 16.680 kDa

CAS No.: 77107-46-1
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder, 16.680 kDa
Appearance: White
Solubility: Soluble in water at 1mg/ml
Merck Reference: 14,1719
MDL No.: MFCD00131869

Activates cyclic phosphodiesterase


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty