Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Human Calcitonin

Supplier: Origen Biomedical, Inc

J63192-EXD J63192-MCR
AAJ63192-EXDEA 279 USD InStock
AAJ63192-EXD AAJ63192-MCR
Human Calcitonin
Proteins and Peptides
Purity: 97+%
CAS No.: 21215-62-3
Molecular Formula: C151H226N40O45S3
Formula Weight: 3417.90
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Merck Reference: 14,1643
MDL No.: MFCD00167520

Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. A carrier peptide that can be used to internalize fusion proteins


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty