H-Arg-Phe-OH acetate, Bachem
Supplier: Frontier Scientific
4002139.0250 4002139.1000
G-1515.0250BAEA 147.06 USD InStock
G-1515.0250BA G-1515.1000BA
H-Arg-Phe-OH acetate, Bachem
Proteins and Peptides
The dipeptide Arg-Phe mimics the haemodynamic effects of FMRF amide. Both peptides increased blood pressure and heart rate in anaesthesized rats by central stimulation of the sympathetic nervous system, RF being approximately 4 fold more potent.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty