H-Glu(Gln-OH)-OH
Supplier: Frontier Scientific
4001245.0250 4001245.1000
G-1930.0250BAEA 441.71 USD InStock
G-1930.0250BA G-1930.1000BA
H-Glu(Gln-OH)-OH
Proteins and Peptides
See also G-4745, H-3996, the branched substrates H-2416, and L-2070, and the product families 'Glutathione-Related Peptides' and 'Ophthalmic Acid'.
γ-Glu-Gln and further kokumi-active γ-glutamyl dipeptides have been isolated from mature Gouda cheese. These branched dipeptides are responsible for the complex taste of the cheese.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty