H-Glu(H-Lys-OH)-OH
Supplier: Frontier Scientific
4001034.0250 4001034.1000
G-1970.0250BAEA 163.38 USD InStock
G-1970.0250BA G-1970.1000BA
H-Glu(H-Lys-OH)-OH
Proteins and Peptides
See also G-4745, H-3996, the branched substrates H-2416, and L-2070, and the product families 'Glutathione-Related Peptides' and 'Ophthalmic Acid'.
This isopeptide bond is formed as a crosslink in proteins in normal, not virus-transformed cells. Additionally, the acceleration of fibrin ε-(γ-glutamyl)lysine crosslinking by red blood cells has been observed.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty