H-Ile-Ile-OH
Supplier: Frontier Scientific
4007632.0001 4007632.0005
G-2405.0001BAEA 1044.85 USD InStock
G-2405.0001BA G-2405.0005BA
H-Ile-Ile-OH
Proteins and Peptides
Contrary to similar non-polar dipeptides as Ile-Leu or Ile-Val, Ile-Ile crystallizes forming hydrophobic columns.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty