Galanin (1-13)-Substance P (5-11) amide
Supplier: Frontier Scientific
4030656.0500 4030656.1000
H-1312.0500BAEA 245.07 USD InStock
H-1312.0500BA H-1312.1000BA
Galanin (1-13)-Substance P (5-11) amide
Proteins and Peptides
The chimaeric peptide galantide is a high-affinity ligand for galanin receptors in membranes from rat hypothalamus, hippocampus, and spinal cord. There it reversibly blocks the neuronal actions of galanin in a dose-dependent manner. The peptide also reverses the galanin-mediated inhibition of insulin release from mouse pancreatic islets. Antagonists such as galantide are therefore of potential importance both as therapeutic agents and in studies on the physiological roles of galanin.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty