Human;Mouse;Rat MCH Trifluoroacetate
Supplier: Frontier Scientific
4025037.0500 4025037.1000
H-1482.0500BAEA 294.14 USD InStock
H-1482.0500BA H-1482.1000BA
Human;Mouse;Rat MCH Trifluoroacetate
Proteins and Peptides
Melanin-concentrating hormone (MCH), originally isolated from salmon pituitary glands, has also been found in the mammalian CNS. In lower vertebrates, salmon MCH alters pigmentation by inducing melanosome aggregation within melanocytes of teleost fish, whereas it causes melanosome dispersion in amphibians and reptiles. The biological role of MCH in higher vertebrates, however, is not yet fully understood, but it may function as a neuromodulator and hypophysiotropic agent in the secretion of α-MSH, ACTH, and GH.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty